Anti-H3K18me1 Rabbit Monoclonal Antibody [clone: RM167]
Supplier: Revmab Biosciences
31-1076-00
103255-480EA 465.23 USD
103255-480
Anti-H3K18me1 Rabbit Monoclonal Antibody [clone: RM167]
Antibodies
This antibody reacts to Histone H3 monomethylated at Lysine 18 (K18me1). No cross reactivity with dimethylated Lysine 18 (K18me2) or trimethylated Lysine 18(K18me3), or other methylations in histone H3.
Western Blot: 1 ug/mL – 2 ug/mL;
ELISA: 0.2 ug/mL - 1 ug/mL;
Multiplex: 0.1 ug/mL – 0.5 ug/mL.