Anti-H3K79me3 Rabbit Monoclonal Antibody [clone: RM157]
Supplier: Revmab Biosciences
31-1053-00
103255-476EA 465.23 USD
103255-476
Anti-H3K79me3 Rabbit Monoclonal Antibody [clone: RM157]
Antibodies
This antibody reacts to Histone H3 trimethylated at Lysine 79 (K79me3). No cross reactivity with monomethylated Lysine 79 (K79me1) or dimethylated Lysine 79 (K79me2), or other methylations in histone H3.
Western Blot: 0.2 ug/mL – 1 ug/mL;
ELISA: 0.1 ug/mL - 0.5 ug/mL;
Multiplex: 0.1 ug/mL – 0.5 ug/mL.