Anti-H3K14me2 Rabbit Monoclonal Antibody [clone: RM165]
Supplier: Revmab Biosciences
31-1060-00
103255-468EA 474.92 USD
103255-468
Anti-H3K14me2 Rabbit Monoclonal Antibody [clone: RM165]
Antibodies
This antibody reacts to Histone H3 dimethylated at Lysine 14 (K14me2). No cross reactivity with monomethylated Lysine 14 (K14me1) or trimethylated Lysine 14 (K14me3), or other methylation in histone H3.
Western Blot: 0.25 ug/mL – 1 ug/mL;
ELISA: 0.2 ug/mL - 1 ug/mL;
Multiplex: 0.1 ug/mL – 0.5 ug/mL.