Anti-H3K9ac Rabbit Monoclonal Antibody [clone: RM161]
Supplier: Revmab Biosciences
31-1054-00
103255-460EA 465.23 USD
103255-460
Anti-H3K9ac Rabbit Monoclonal Antibody [clone: RM161]
Antibodies
This antibody reacts to Histone H3 acetylated at Lysine 9 (K9ac). No cross reactivity with other acetylated Lysines in histone H3.
Western Blot: 0.25 ug/mL - 1 ug/mL;
ICC: 0.5 ug/mL - 2 ug/mL;
ChIP: 2 ug/mL-10 ug/mL;
IHC: 0.1 ug/mL – 1 ug/mL;
ELISA: 0.2 ug/mL - 1 ug/mL;
Multiplex: 0.05 ug/mL – 0.5 ug/mL.