Anti-H3K36ac Rabbit Monoclonal Antibody [clone: RM154]
Supplier: Revmab Biosciences
31-1050-00
103255-454EA 474.92 USD
103255-454
Anti-H3K36ac Rabbit Monoclonal Antibody [clone: RM154]
Antibodies
This antibody reacts to Histone H3 acetylated at Lysine 36 (K36ac), which is not affected by the phosphorylation of neighboring Ser31. No cross reactivity with other acetylated Lysines in histone H3.
Western Blot: 0.5 ug/mL - 2 ug/mL;
ICC: 0.5 ug/mL - 2 ug/mL;
ELISA: 0.2 ug/mL - 1 ug/mL;
Multiplex: 0.1 ug/mL – 0.5 ug/mL.