Anti-H3K4me3 Rabbit Monoclonal Antibody [clone: RM137]
Supplier: Revmab Biosciences
31-1039-00
103255-448EA 465.23 USD
103255-448
Anti-H3K4me3 Rabbit Monoclonal Antibody [clone: RM137]
Antibodies
This antibody reacts to Histone H3 trimethylated at Lysine 4 (K4me3). No cross reactivity with monomethylated Lysine 4 (K4me1) or dimethylated Lysine 4 (K4me3), or other methylation in histone H3.
Western Blot: 0.2 ug/mL – 1 ug/mL;
ELISA: 0.1 ug/mL – 0.5 ug/mL;
Multiplex: 0.1 ug/mL – 0.5 ug/mL.