Anti-H3K4me1 Rabbit Monoclonal Antibody [clone: RM140]
Supplier: Revmab Biosciences
31-1046-00
103255-444EA 474.92 USD
103255-444
Anti-H3K4me1 Rabbit Monoclonal Antibody [clone: RM140]
Antibodies
This antibody reacts to Histone H3 monomethylated at Lysine 4 (K4me1). No cross reactivity with dimethylated Lysine 4 (K4me2) or trimethylated Lysine 4 (K4me3), or other methylations in histone H3
Western Blot: 0.2 ug/mL – 1 ug/mL;
ChIP: 2 ug/mL- 10 ug/mL;
ICC: 1 ug/mL - 2 ug/mL;
ELISA: 0.2 ug/mL - 1 ug/mL;
Multiplex: 0.1 ug/mL – 0.5 ug/mL.