Anti-TUBA Rabbit Monoclonal Antibody [clone: RM113]
Supplier: Revmab Biosciences
31-1016-00
103255-436EA 382.31 USD
103255-436
Anti-TUBA Rabbit Monoclonal Antibody [clone: RM113]
Antibodies
This antibody reacts to α-Tubulin, including Tubulin alpha-1A chain and Tubulin alpha-1B chain.
Western Blot (WB): 1:1000 dilution;
Immunoprecipitation (IP): 1:200 dilution;
Chromatin IP (ChIP): 1:200 dilution;
Immunocytochemistry (ICC): 1:200 dilution;
Immunohistochemistry (IHC): 1:200 dilution.