Anti-Tyr Rabbit Monoclonal Antibody [clone: RM111]
Supplier: Revmab Biosciences
32-1011-00
103255-434EA 431.85 USD
103255-434
Anti-Tyr Rabbit Monoclonal Antibody [clone: RM111]
Antibodies
This antibody reacts to tyrosine-phosphorylated proteins. No cross reactivity with nonphosphorylated tyrosine, phosphoserine, and phosphorthreonine
Western Blot (WB): 1:1000 -1:5000 dilution;
Immunoprecipitation (IP): 1:200-1:1000 dilution;
Chromatin IP (ChIP): 1:200-1:1000 dilution;
Immunocytochemistry (ICC): 1:200-1:500 dilution;
Immunohistochemistry (IHC): 1:200-1:500 dilution.