Anti-Thr Rabbit Monoclonal Antibody [clone: RM102]
Supplier: Revmab Biosciences
31-1009-00
103255-230EA 440.46 USD
103255-230
Anti-Thr Rabbit Monoclonal Antibody [clone: RM102]
Antibodies
This antibody reacts threonine-phosphorylated proteins. No cross reactivity with nonphosphorylated threonine, phosphoserine, and phosphotyrosine. It shows slight cross-reactivity with a few phospho-serine-containing peptides.
Western Blot (WB): 1:500 -1:2000 dilution;
Immunoprecipitation (IP): 1:100-1:500 dilution;
Chromatin IP (ChIP): 1:100-1:500 dilution;
Immunocytochemistry (ICC): 1:100-1:500 dilution;
Immunohistochemistry (IHC): 1:100-1:500 dilution.