Anti-Lys Rabbit Monoclonal Antibody [clone: RM101]
Supplier: Revmab Biosciences
32-1012-00
103255-228EA 440.46 USD
103255-228
Anti-Lys Rabbit Monoclonal Antibody [clone: RM101]
Antibodies
This antibody reacts to lysine-acetylated proteins. No cross reactivity with nonacetylated lysine, and lysine with other modification.
Western Blot (WB): 1:500 -1:2000 dilution;
Immunoprecipitation (IP): 1:100-1:500 dilution;
Chromatin IP (ChIP): 1:100-1:500 dilution;
Immunocytochemistry (ICC): 1:100-1:500 dilution;
Immunohistochemistry (IHC): 1:100-1:500 dilution.