Anti-IgG Goat Monoclonal Antibody [clone: RMG05] (Biotin)
Supplier: Revmab Biosciences
41-1043-02
103255-428EA 438.31 USD
103255-428
Anti-IgG Goat Monoclonal Antibody [clone: RMG05] (Biotin)
Antibodies
This antibody reacts to the Fab region of Mouse IgG, including IgG1, IgG2a, IgG2b, and IgG3. No cross reactivity with IgM, IgA, IgE, human IgG, rat IgG, or rabbit IgG.
ELISA: 0.05ug/mL – 1ug/mL;
ICC, IHC: 0.5ug/mL – 2ug/mL.