Anti-IgA1 Rabbit Monoclonal Antibody [clone: RM124] (Biotin)
31-1026-02
103255-406EA 284.62 USD
103255-406
Anti-IgA1 Rabbit Monoclonal Antibody [clone: RM124] (Biotin)
This antibody reacts to human IgA1, and very slightly cross reacts with IgA2. No cross reactivity with human IgG, IgM, IgD, or IgE.
ELISA: 50ng/well – 200ng/well (for Capture);
0.05ug/mL – 0.2ug/mL (for Detection);
Immunocytochemistry (ICC): 0.5ug/mL-2ug/mL;
Immunohistochemistry (IHC): 0.1ug/mL-1ug/mL.