Anti-IgG2b Rabbit Monoclonal Antibody [clone: RM108] (Biotin)
31-1004-02
103255-386EA 284.62 USD
103255-386
Anti-IgG2b Rabbit Monoclonal Antibody [clone: RM108] (Biotin)
This antibody reacts to the Fc region of mouse IgG2b. No cross reactivity with mouse IgG1, IgG2a, IgG3, IgM, IgA, IgE, human IgG, rat IgG, or goat IgG.
ELISA: 0.005 ug/mL – 0.2 ug/mL;
Immunocytochemistry (ICC): 0.5 ug/mL-2 ug/mL;
Immunohistochemistry (IHC): 0.5 ug/mL-2 ug/mL;
Western Blot (WB): 0.1 ug/mL-0.5 ug/mL.