Anti-IgG4 Rabbit Monoclonal Antibody [clone: RM120]
31-1022-00
103255-370EA 338.46 USD
103255-370
Anti-IgG4 Rabbit Monoclonal Antibody [clone: RM120]
This antibody reacts to the heavy chain of human IgG4. No cross reactivity with human IgG1, IgG2, IgG3, IgM, IgA, IgD, IgE, mouse IgG, rat IgG, or goat IgG.
ELISA: 50ng/well – 200ng/well (for Capture);
0.05ug/mL – 0.2ug/mL (for Detection);
Western Blot (WB): 0.5 ug/mL – 2 ug/mL;
Immunocytochemistry (ICC): 1 ug/mL-10ug/mL;
Immunohistochemistry (IHC): 1 ug/mL-10ug/mL.