Anti-IgG Goat Monoclonal Antibody [clone: RMG07]
Supplier: Revmab Biosciences
41-1045-00
103255-214EA 429.69 USD
103255-214
Anti-IgG Goat Monoclonal Antibody [clone: RMG07]
Antibodies
This antibody reacts to Mouse IgG, including IgG1, IgG2a, IgG2b, and IgG3. No cross reactivity with IgM, IgA, IgE, human IgG, rat IgG, or rabbit IgG.
ELISA: 0.05ug/mL – 1ug/mL;
ICC, IHC: 0.5ug/mL – 2ug/mL.