Anti-IgG Goat Monoclonal Antibody [clone: RMG02]
Supplier: Revmab Biosciences
41-1015-00
103255-344EA 429.69 USD
103255-344
Anti-IgG Goat Monoclonal Antibody [clone: RMG02]
Antibodies
This antibody reacts to the Fc region of Rabbit IgG. No cross reactivity with human IgG, rat IgG, or mouse IgG.
ELISA: 0.01 ug/mL – 0.5 ug/mL;
Immunocytochemistry (ICC): 0.5 ug/mL-2 ug/mL;
Immunohistochemistry (IHC): 0.5 ug/mL-2 ug/mL;
Western Blot (WB) (nonreduced): 0.1 ug/mL-0.5 ug/mL.