Anti-IgA Rabbit Monoclonal Antibody [clone: RM220]
31-1099-00
103255-200EA 323.08 USD
103255-200
Anti-IgA Rabbit Monoclonal Antibody [clone: RM220]
This antibody specifically reacts to mouse IgA. No cross reactivity with mouse IgG1, IgG2a, IgG2b, IgG2c, IgG3, IgM, IgE, or human IgA.
ELISA: 0.005 ug/mL – 0.2 ug/mL;
Immunocytochemistry (ICC): 0.5 ug/mL-2 ug/mL;
Immunohistochemistry (IHC): 0.5 ug/mL-2 ug/mL;
Western Blot (WB): 0.5 ug/mL-2 ug/mL.