Orion™ 131S Basic Waterproof Conductivity Meter, Thermo Scientific
0131S2 0131S3 0131S0
23197-858EA 2880.42 USD
23197-858 34105-228 34105-230
Orion™ 131S Basic Waterproof Conductivity Meter, Thermo Scientific
Measure conductivity, salinity, total dissolved solids (TDS), and temperature using the Orion™ 131S Intrinsically Safe (US) Portable Conductivity Meter.
  • Calibration via Thermo Scientific AUTO-CAL™ and cell constant adjustment method
  • Compatibility with two-cell or four-cell electrodes
  • Temperature compensation options, including automatic or manual linear function, and non-linear function for ultrapure or natural water
  • Easy-to-use interface cell status and self-diagnostic testing
  • Custom LCD
  • Safe for use in a non-sparking environment
Measure conductivity, salinity, total dissolved solids (TDS), and temperature in environments where non-sparking equipment is required using the Orion™ 131S Intrinsically Safe (US) Portable Conductivity Meter. Intrinsically safe approval rating by Factory Mutual (FM) for US only.
Certifications: CE, FM (IS, Class I Div 1 Group A, B, C, D, T4), Ex (Intrinsically Safe)