New Brunswick Galaxy® 48 Series CO2 Incubators, Eppendorf
CO48S1200000
89171-972EA 8553.98 USD
89171-972
New Brunswick Galaxy® 48 Series CO2 Incubators, Eppendorf
Galaxy 48 S incubator is specifically designed for smaller laboratories and applications requiring a higher level of isolation.
  • Small footprint with mid-sized capacity
  • Seamless chamber and fanless design
  • RS-232 port and 25 mm access port
  • User-friendly LED display
  • IR sensor and low gas consumption
Galaxy 48 S incubator is an entry-level model, offering the same superior performance as 48 R models, but with an easy-to-use keypad and LED display.
This provide many of the same standard features as the 170 l systems to provide sophisticated performance in a smaller package and include a viewing window on the outside door.