Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Attached Lid Container, Akro-Mils

Supplier: AKRO-MILS
39-120 39-085 39-170
Attached Lid Containers (ALCs) are reusable storage containers ideal for use in order picking, closed-loop distribution, and storage applications.

  • Textured Bottoms
  • Handle Grips
  • Padlock Eye
  • Reinforced Steel Hinge Pins
  • Reusable
  • Twelve Gallon Capacity

Attached lids snap securely closed to protect contents from dust or damage. These industrial shipping containers stack for maximized storage and nest when empty to save space.

Textured bottoms provide a sure grip on conveyor belts. Strong molded-in handle grips are ergonomically designed for easy lifting and carrying. Padlock eye provides a security option. Reinforced steel hinge pins provide years of smooth lid operation.

Ordering information: Eight, Twelve and Seventeen Gal. Capacity



 

ORDER

Expand All / Collapse All
Exterior Height Exterior Width Top Inside Dimensions Exterior Length Bottom Interior Dimensions Inside Height Supplier No. VWR Catalog Number Unit Price Quantity
121/2" 15" 141/2" 211/2" 131/4" 12" 39-120 47751-095 Case of 6 Retrieving
17" 15" 141/2" 211/2" 131/4" 167/8" 39-170 47751-097 Case of 3 Retrieving
9" 15" 141/2" 211/2" 131/4" 85/8" 39-085 47751-093 Case of 6 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty