Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Analytical Balance, Model XPE206DR, METTLER TOLEDO®

Supplier: Mettler Toledo
XPE
30132913
METTLER TOLEDO XPE analytical balances provide outstanding performance in analytical weighing and support the highest requirements for safety, efficiency and ease of compliance.

  • Capacity 81g/220g
  • Readability 5mg/0.01mg

Thanks to excellent repeatability, XPE analytical balances offer you the smallest minimum weight down to 10mg. Quality management features, such as the innovative StatusLight and patented StaticDetect technology, take the worry out of weighing and provide you with a high level of trust in your results. With a wide range of accessories available and multiple connectivity options, XPE Balances grow with your needs. You can enjoy a wealth of weighing possibilities for years to come. Designed, engineered and manufactured in Switzerland for outstanding quality you can trust. Secure the future though simple and immediate upgrading to automated powder and liquid dosing, to improve security, user safety and ensure accurate dosing results.

Certifications: UL listed and CSA or CUL (North America only) certified.

Ordering information: Service packages are offered to assure continual reliability and maintain optimum balance performance. Proper installation and initial calibration are the key to ensuring the balance performs properly from day one. A METTLER TOLEDO technician performs installation qualification and operational qualification (IQ/OQ). Contact your representative for Service package details.




 

ORDER

Expand All / Collapse All
Model Weighing Capacity Linearity Pan/Platform Size Readability Stabilization Time Repeatabilty Supplier No. VWR Catalog Number Unit Price Quantity
XPE206DR 81 g/220 g 0.10 mg 31/16 x 27/8" 0.005 mg/0.01 mg 2.5 s/3.5 s 0.015 mg (@ 5% load) 30132913 75792-194 Each Retrieving
UL Listed CSA Certified
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Communication Interface RS-323, USB Host, USB Device
Dimensions 21 x 15 x 18"
Voltage 100–240VAC, 0.8A, 50/60Hz, 12V DC 2.5A

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty