Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Speedglas™ 9100 Series Welding Helmet, 3M™

Supplier: 3M OCCUPATIONAL HEALTH
Speedglas™
51141563596 51141563633 51141563602 51141563657 51141563619
This hard hat and welding helmet system allows the welding helmet to nest over a hard hat when raised

  • All Speedglas™ 9100 Series Welding Helmets feature dark shades 5, 8 – 13
  • Battery type for all helmets is CR2032
  • Helmets are ideally suited for MIG welding, stick welding, and TIG welding processes

In addition, users can easily detach the welding helmet while a hard hat remains in place for non-welding tasks.



 

ORDER

Expand All / Collapse All
Description Color Supplier No. VWR Catalog Number Unit Price Quantity
Speedglas™ 9100 Series Welding Helmet, with 1.8 x 3.7" ADF Viewing Area and 2800 hr. Battery Life Black 51141563596 10204-890 Case of 1 Retrieving
Speedglas™ 9100 Series Welding Helmet, with 2.1 x 4.2" ADF Viewing Area and 2500 hr. Battery Life Black 51141563602 10204-940 Case of 1 Retrieving
Speedglas™ 9100 Series Welding Helmet, with 2.8 x 4.2" ADF Viewing Area and 2000 hr. Battery Life Black 51141563619 10204-942 Case of 1 Retrieving

Replacement Welding Helmet
Description Color Supplier No. VWR Catalog Number Unit Price Quantity
Speedglas™ 9100 Series Replacement Welding Helmet, with Rail Black 51141563633 10204-894 Case of 1 Retrieving
Speedglas™ 9100 Series Replacement Welding Helmet, without Rail Black 51141563657 10204-898 Case of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

ACCESSORIES

Expand All / Collapse All

Accessories
Description Details For Use With Color Supplier No. VWR Catalog Number Unit Price Quantity
3M™ High Temperature Welding Shield L-184 4.5 x 5.25" Headgear L-505, L-705 and L-905 Silver L-184 67101-214 Case of 1 Retrieving
3M™ Lens Gasket 061-35-01R01 Breathe Easy™ and Supplied Air Welding Helmets Black 061-35-01R01 67100-038 Each Retrieving
3M™ Speedglas™ Earmuff Expander Bar 9100 QR, 06-0118-00QR Includes earmuff bar and 2 outside pivot cover plates Speedglas™ Welding Helmet 9100 QR 51141563701 10204-948 Case of 1 Retrieving
3M™ Speedglas™ Pivot Cover Plate Kit 06-0116-00QR Includes 2 inside and 2 outside pivot cover plates Speedglas™ Welding Helmet Series QR 51141563695 10204-946 Case of 1 Retrieving
3M™ Welding Gasket L-142-2 4.5 x 5.25" Lens stacks Black L-142-2 67101-188 Case of 2 Retrieving
3M™ Welding Knob and Pivot Kit L-146 Includes Welding Knob L-145 and pivot post Welding Faceshield and Combination Faceshield Black L-146 67101-192 Case of 1 Retrieving
3M™ Welding Knob L-145-10 Secures cam mechanism Welding Shields L-150, L-151 and Combination Faceshields L-153, L-154, L-184, and L-185 Black L-145-10 67101-190 Case of 10 Retrieving
3M™ Welding Lens Retainer Clip L-147-2e Lens stacks Metal L-147-2 67101-194 Case of 2 Retrieving
3M™ Welding Shield L-150 4.5 x 5.25" Bumpcap L-503 and Hardhat L-703 Black L-150 67101-198 Case of 1 Retrieving
3M™ Welding Shield L-153 4.5 x 5.25" Bumpcap L-505, Hardhat L-705, and Helmet L-905 Black L-153 67101-202 Case of 1 Retrieving
Speedglas™ Quick Release Rail - - Black 51141563671 10204-902 Case of 1 Retrieving
Speedglas™ Rail Retention Parts, 3M™ - - - 51141563688 10204-944 Case of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty