Would you like to log in again?
Calibration/Liquid Measuring Flasks, TC U.S. Series, Wilmad-LabGlass
Supplier: WILMAD/LABGLASS
LG-4310-108 LG-4310-104 LG-4310-106 LG-4310-100 LG-4310-112 LG-4310-102 LG-4310-114 LG-4310-110
Intended for checking automatic fill volumes, the borosilicate glass cylinder and flasks are calibrated "to contain" (TC) at 68°F
The complete set includes two cases – one with a 2 oz. graduated cylinder and larger sized flasks consisting of 1 gill, 1/2 pint, 1 pint, 1 quart, ½ gal. The 1 gal flask comes in a case of its own.
ORDER
Expand All / Collapse All
Description Nominal Capacity Graduations Tolerance Supplier No. VWR Catalog Number Unit Price Quantity
Cylinder 2 fl. oz. ±5/0.31 min./mL (at Nominal);
±5/0.31 min./mL (within Graduated Portion)
LG-4310-102 14201-218 Each Retrieving
Flask 1 gill 1/2 x 1/4 fl. drams (Graduated Above/Below Nominal x Subdivisions) ±3/0.18 min./mL (at Nominal);
±1/0.06 min./mL (within Graduated Portion)
LG-4310-104 14212-736 Each Retrieving
Flask 1/2 pint 1 x 1/4 fl. drams (Graduated Above/Below Nominal x Subdivisions) ±4/0.25 min./mL (at Nominal);
±1.5/0.09 min./mL (within Graduated Portion)
LG-4310-106 14200-578 Each Retrieving
Flask 1 pint 2 x 1/2 fl. drams (Graduated Above/Below Nominal x Subdivisions) ±7/0.43 min./mL (at Nominal);
±3/0.18 min./mL (within Graduated Portion)
LG-4310-108 14201-084 Each Retrieving
Flask 1 quart 4 x 1 fl. drams (Graduated Above/Below Nominal x Subdivisions) ±12/0.74 min./mL (at Nominal);
±5/0.31 min./mL (within Graduated Portion)
LG-4310-110 14201-076 Each Retrieving
Flask 1/2 gal. 6 x 1 fl. drams (Graduated Above/Below Nominal x Subdivisions) ±16/0.99 min./mL (at Nominal);
±5/0.31 min./mL (within Graduated Portion)
LG-4310-112 14200-710 Each Retrieving
Flask 1 gal. 8 x 1 fl. drams (Graduated Above/Below Nominal x Subdivisions) ±20/1.23 min./mL (at Nominal);
±5/0.31 min./mL (within Graduated Portion)
LG-4310-114 14200-692 Each Retrieving
Complete Set 2 fl. oz., 1 gill, 1/2 pint, 1 pint, 1 quart, 1/2 gal, and 1 gal LG-4310-100 14200-958 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Q and A
Product Questions & Answers
Sort by: Most Helpful | Newest | Oldest
Items Per Page: 5  10  20 
2  3  4  Next
Can't find an answer to your question?  Ask us a question
Ask us a question:
Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question
Q&A Guidelines & Tips
If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.
When writing your question or answer, please follow these guidelines:
  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English
All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s
We reserve the right not to post your question if it contains any of the following
  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty