Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PCR Reagents, GenScript

Supplier: Genscript
C01580-250ML C01577-10ML C01578-1ML C01577-250ML C01578-50ML C01582-50ML C01578-250ML C01580-10ML C01577-50ML C01580-50ML C01579-50ML C01579-250ML C01579-1ML C01580-1ML C01578-10ML C01582-10ML C01582-1ML C01579-10ML C01577-1ML C01582-250ML
High throughput PCR automation, offering cost-effective solution and satisfactory results.

Applied for PCR, 3'A-tailing of blunt ends, Primer extension and DNA sequencing.




 

ORDER

Expand All / Collapse All
Description Size Supplier No. VWR Catalog Number Unit Price Quantity
DATP, 100 mM, ≥99% HPLC 250 mL (250.0 mL/bottle) C01577-250ML 95044-186 Each (250ml) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

DATP, 100 mM, ≥99% HPLC 1 mL (1.0 mL/vial) C01577-1ML 95044-180 Each (1ml) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

DATP, 100 mM, ≥99% HPLC 50 mL (2x25 mL) C01577-50ML 95044-184 Each (50ml) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

DATP, 100 mM, ≥99% HPLC 10 mL (10.0 mL/vial) C01577-10ML 95044-182 Each (10ml) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

DCTP, 100 mM, ≥99% HPLC 250 mL (250.0 mL/bottle) C01579-250ML 95044-202 Each (250ml) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

DCTP, 100 mM, ≥99% HPLC 10 mL (10.0 mL/vial) C01579-10ML 95044-198 Each (10ml) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

DCTP, 100 mM, ≥99% HPLC 1 mL (1.0 mL/vial) C01579-1ML 95044-196 Each (1ml) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

DCTP, 100 mM, ≥99% HPLC 50 mL (2x25 mL) C01579-50ML 95044-200 Each (50ml) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

DGTP, 100 mM, ≥99% HPLC 50 mL (2x25 mL) C01578-50ML 95044-192 Each (50ml) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

DGTP, 100 mM, ≥99% HPLC 10 mL (10.0 mL/vial) C01578-10ML 95044-190 Each (10ml) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

DGTP, 100 mM, ≥99% HPLC 1 mL (1.0 mL/vial) C01578-1ML 95044-188 Each (1ml) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

DGTP, 100 mM, ≥99% HPLC 250 mL (250.0 mL/bottle) C01578-250ML 95044-194 Each (250ml) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

DNTP, 10 mM each 50 mL (2x25 mL) C01582-50ML 95040-882 Each Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

View MSDS SDS
DNTP, 10 mM each 1 mL (1.0 mL/vial) C01582-1ML 95040-878 Each Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

View MSDS SDS
DNTP, 10 mM each 250 mL (250.0 mL/bottle) C01582-250ML 95040-884 Each Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

View MSDS SDS
DNTP, 10 mM each 10 mL (10.0 mL/vial) C01582-10ML 95040-880 Each Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

View MSDS SDS
DTTP, 100 mM, ≥99% HPLC 10 mL (10.0 mL/vial) C01580-10ML 95044-206 Each (10ml) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

DTTP, 100 mM, ≥99% HPLC 250 mL (250.0 mL/bottle) C01580-250ML 95044-210 Each (250ml) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

DTTP, 100 mM, ≥99% HPLC 1 mL (1.0 mL/vial) C01580-1ML 95044-204 Each (1ml) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

DTTP, 100 mM, ≥99% HPLC 50 mL (2x25 mL) C01580-50ML 95044-208 Each (50ml) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty