Difco™ Thermoacidurans Agar, BD Biosciences
Supplier: BD Biosciences
230310 230310
90002-060EA 458.87 USD
CA90002-060 90002-060
Difco™ Thermoacidurans Agar, BD Biosciences
Media for Microbiology Additives and Supplements for Microbiology
Thermoacidurans Agar is used for isolating and cultivating Bacillus coagulans (Bacillus thermoacidurans) from foods.
  • Peptones provide carbon and nitrogen
  • Agar is the solidifying agent
  • Dehydrated appearance: Light tan, free-flowing
Thermoacidurans Agar can also be used to isolate mesophilic spore-forming anaerobes from foods. These microorganisms tolerate high heat, grow in the absence of oxygen, and grow over the range of temperatures used in canned and processed foods. They are of primary importance in spoilage of low-acid foods packed in hermetically sealed containers.