Difco™ Staphylococcus Medium 110, BD Biosciences
Supplier: BD Life Sciences
229730 229730
90002-042EA 207.99 USD
CA90002-042 90002-042
Difco™ Staphylococcus Medium 110, BD Biosciences
Media for Microbiology Dehydrated Media for Microbiology
Staph 110 Medium is a solid medium recommended for use in qualitative procedures for selective isolation of Staphylococcus species
  • Dehydrated appearance: Beige, free-flowing
  • Peptone provides carbon, nitrogen, vitamins, and minerals
  • Sodium chloride inhibits most bacteria other than staphylococci
  • Agar is the solidifying agent
Gelatin is included for testing liquefaction.