BBL™ SIM Medium, BD Biosciences
Supplier: BD Biosciences
211578 211578
90000-232EA 308.66 USD
CA90000-232 90000-232
BBL™ SIM Medium, BD Biosciences
Media for Microbiology Dehydrated Media for Microbiology
Sulfide Indole Motility (SIM) Medium is used for the determination of sulfide production, indole formation, and motility of enteric microorganisms.
  • Dehydrated appearance: Beige, free-flowing
Hydrogen sulfide production, indole formation and motility are distinguishing characteristics which aid in the identification of the Enterobacteriaceae, especially Salmonella and Shigella. SIM Medium, therefore, is useful in the process of identification of enteric pathogens.