Difco™ Sabouraud Maltose Broth, BD
Supplier: BD Diagnostics
242910 242910
90003-286EA 452.1 USD
90003-286 CA90003-286
Difco™ Sabouraud Maltose Broth, BD
Media for Microbiology Dehydrated Media for Microbiology
Sabouraud Maltose Agar is used for the cultivation of yeasts, molds, and aciduric microorganisms.
  • Peptone provides nitrogen, vitamins, minerals, amino acids, and growth factors
  • Low pH promotes the growth of fungi
The addition of antimicrobics further increases the selectivity of the medium.