Would you like to log in again?
Uvex Ambient OTG® Lens, Honeywell Safety
Supplier: HONEYWELL SAFETY PRODUCTS USA INC.
S3160 S3162 S3150 S3152 S3163 S3165 S3150X S3152X S3150D
Uvex Ambient eyewear provides over-the-glass coverage, offering workers full protection while wearing their own prescription eyewear
  • Lightweight, stylish and comfortable
  • Exceptional peripheral vision and coverage
  • Wraparound uni-lens design
  • Soft, flexible nasal fingers conform to a variety of facial profiles and minimize slippage
  • Adjustable and bendable, wire-core temples offer complete flexibility with secure, customized fit
  • Economical and easy lens replacement allows multiple tints and extends the life of the eyewear
Ideal for use in agriculture, food services, government, manufacturing, medical, mining, municipal services, oil and gas, pharmaceutical and welding industries.
Certifications: Meets ANSI Z87.1-2003 (High Impact) standard and certified to the requirements of CSA Z94.3.
ORDER
Expand All / Collapse All
Frame Material Description Size Frame Color Lens Coating Lens Tint Lens Material Temple Type Supplier No. VWR Catalog Number Unit Price Quantity
Polycarbonate Uvex Ambient OTG Medium Blue Ultra-dura Anti-scratch Clear Polycarbonate Flexible Wire Core S3160 89095-493 Each Retrieving
Pack of 10 Retrieving
Polycarbonate Uvex Ambient OTG Medium Blue Ultra-dura Anti-scratch Gray Polycarbonate Flexible Wire Core S3162 89095-497 Each Retrieving
Polycarbonate Uvex Ambient OTG Medium Blue Ultra-dura Anti-scratch Shade 5.0 Infra-dura Polycarbonate Flexible Wire Core S3165 89095-500 Each Retrieving
Pack of 10 Retrieving
Polycarbonate Uvex Ambient OTG Large Blue Ultra-dura Anti-scratch Clear Polycarbonate Flexible Wire Core S3150 89095-475 Each Retrieving
Pack of 10 Retrieving
Polycarbonate Uvex Ambient OTG Large Blue UV Extreme Anti-fog Clear Polycarbonate Flexible Wire Core S3150X 89095-477 Each Retrieving
Polycarbonate Uvex Ambient OTG Large Blue Dura-streme Dual (Anti-Fog/Anti-Scratch) Clear Polycarbonate Flexible Wire Core S3150D 89095-478 Each Retrieving
Pack of 10 Retrieving
Polycarbonate Uvex Ambient OTG Large Blue Ultra-dura Anti-scratch Gray Polycarbonate Flexible Wire Core S3152 89095-485 Each Retrieving
Polycarbonate Uvex Ambient OTG Large Blue UV Extreme Anti-fog Gray Polycarbonate Flexible Wire Core S3152X 89095-486 Each Retrieving
Pack of 10 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Q and A
Product Questions & Answers
Sort by: Most Helpful | Newest | Oldest
Items Per Page: 5  10  20 
2  3  4  Next
Can't find an answer to your question?  Ask us a question
Ask us a question:
Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question
Q&A Guidelines & Tips
If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.
When writing your question or answer, please follow these guidelines:
  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English
All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s
We reserve the right not to post your question if it contains any of the following
  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty