Would you like to log in again?
BioStor™ Vials with Screw Cap, National Scientific Supply
Supplier: NATIONAL SCIENTIFIC SUPPLY CO
BS20NA-PS BC20NA-PS BC20AS-PS BC20NA-BP BC20AS-BP
These screw cap vials are widely used for sample collection and processing, centrifugation, long-term storage, and packing and shipping of valuable biological reagents.
  • Easy to handle
  • Minimizes the risk of contamination
  • O-rings are uniformly compressed for unparalleled secure sealing
  • Deep inner skirt on cap is reinforced to withstand high temperature
  • Uniform, thick walled construction with reinforced support collar
  • Centrifugation to 17,000×g
Cap has a 3mm syringe port which is pierceable for access of hazardous materials or fraction collecting without opening.
ORDER
Expand All / Collapse All
Description Capacity Cap Color Sterility Temperature Range Supplier No. VWR Catalog Number Unit Price Quantity
BioStor™ Vials with Screw Cap, Conical 2 mL Natural Sterile –195 to 121°C BC20NA-PS 66006-830 Case of 500 Retrieving
BioStor™ Vials with Screw Cap, Conical 2 mL Assorted Nonsterile –195 to 121°C BC20AS-BP 66006-805 Case of 1,000 Retrieving
BioStor™ Vials with Screw Cap, Conical 2 mL Natural Nonsterile –195 to 121°C BC20NA-BP 66006-804 Case of 1,000 Retrieving
BioStor™ Vials with Screw Cap, Conical 2 mL Assorted Sterile –195 to 121°C BC20AS-PS 66006-831 Case of 500 Retrieving
BioStor™ Vials with Screw Cap, Skirted 2 mL Natural Sterile –195 to 121°C BS20NA-PS 66006-843 Case of 500 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Q and A
Product Questions & Answers
Sort by: Most Helpful | Newest | Oldest
Items Per Page: 5  10  20 
2  3  4  Next
Can't find an answer to your question?  Ask us a question
Ask us a question:
Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question
Q&A Guidelines & Tips
If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.
When writing your question or answer, please follow these guidelines:
  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English
All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s
We reserve the right not to post your question if it contains any of the following
  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty