Human Plasma, Equitech Enterprises
Supplier: Equitech-Bio
SDFDPHP-0100
103219-988EA 193.81 USD
103219-988
Human Plasma, Equitech Enterprises
Plasma
Equitech Enterprise’s human serum products are collected in donor centers located in the United States.
  • Sterile-filtered to 0.2 micron
  • Sterility tested x 14 days
  • Each unit is tested for Anti-HIV 1/2, Anti-HCV, HBs Ag and RPR.
Charcoal Stripping reduces the concentration of steroid hormones in serum including as estradiol, progesterone, cortisol, testosterone T3 and T4.