Cynomolgus monkey Albumin (from Serum)
Supplier: Equitech Bio
CMSA62-0050 CMSA62-0100
103219-930EA 1910.09 USD
103219-930 103219-928
Cynomolgus monkey Albumin (from Serum)
Prepared from cynomolgus monkey serum by heat-shock fractionation.
  • Purity: ≥ 96%
  • IgG level: not detected
Equitech-Bio offers a wide range of heat-shock and proprietary Cohn method processed animal serum albumins. With our high purity albumins, researchers can formulate and customize their own specialized media for specific applications. Prepared from Cynomolgus Monkey serum by heat shock method.