Coenzyme Q10 USP dietary supplement
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
(all-E)-2-(3,7,11,15,19,23,27,31,35,39-Decamethyl-2,6,10,14,18,22,26,30,34,38-tetracontadecaenyl)-5,6-dimethoxy-3-methyl-2,5-cyclohexadiene-1,4-dione
, Ubidecarenone
, Ubiquinone 10
U1310-1KGBL
U1310-100GMPL
U1310-25GM
U1310-1GM
U1310-5GM
700004-192EA
210
USD
700004-192
75812-586
700004-196
700004-194
700004-198
Coenzyme Q10 USP dietary supplement
Coenzyme Q10
Ubidecarenone, USP Dietary Supplement.
All Spectrum Chemical USP Dietary Supplement grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).
Formula:
C₅₉H₉₀O₄ MW: 863.37 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00042919 CAS Number: 303-98-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...