Rabbit Plasma, Equitech- Bio
Supplier: Equitech-Bio
SRPH35-0100 SRPH35-0500 RPE07-0500 RPE07-0100 RPC07-0500 RPH07-0100 RPC07-0100 SRPC35-0500 RPH07-0500 SRPE35-0500 SRPE35-0100
103219-784EA 120.33 USD
103219-784 103219-774 103219-786 103219-776 103219-780 103219-792 103219-782 103219-794 103219-790 103219-788 103219-778
Rabbit Plasma, Equitech- Bio
Collected from 8-12 week-old New Zealand or Californian rabbits
  • Sterility tested
  • Lot to lot consistency
  • Low hemoglobin levels
Raw plasma is non-sterile. Low hemoglobin levels. Physiochemical constituents measured to ensure lot-to-lot consistency. Sterile-Filtered plasma is sterility tested and sterile-filtered to 0.2 micron absolute. Subject to CITES regulations.
Aseptically collected and processed to minimize hemolytic activity.