Rhesus Monkey Plasma, Equitech- Bio, Inc.
Supplier: Equitech-Bio
RMPC07-0050 SRMPH35-0050 RMPE07-0050
103219-114EA 916.83 USD
103219-114 103219-116 103219-112
Rhesus Monkey Plasma, Equitech- Bio, Inc.
Collected from sexually mature, donor, rhesus monkeys that are free of common parasites.
  • Sterility tested
  • Lot to lot consistency
  • Low hemoglobin levels
Raw plasma is non-sterile. Low hemoglobin levels. Physiochemical constituents measured to ensure lot-to-lot consistency. Sterile-Filtered plasma is sterility tested and sterile-filtered to 0.2 micron absolute. Subject to CITES regulations.