New Born Calf Serum, Equitech- Bio
Supplier: Equitech-Bio
SNBU30-0100 SNBU30-0100HI NBU05-0500 SNBU30-0500HI
103219-014EA 37.21 USD
103219-014 103219-004 103219-010 103219-008
New Born Calf Serum, Equitech- Bio
Sera (Serum)
A cost-effective serum for general cell growth requirements.
  • Sterility tested
  • Sterile filtered to 0.2 micron absolute
  • Low hemoglobin levels
  • Physiochemical constituents measured to ensure lot to lot consistency
  • Endotoxin testing available
  • Mycoplasma testing available
  • Heat inactivation is available if desired
Collected from animals of U.S. origin. All serum is collected from calves aged 10 days old or less. Raw, non-sterile with low hemoglobin levels. Physiochemical constituents measured to ensure lot to lot consistency. Endotoxin and mycoplasma testing available.
Certifications: Country of origin: USA
Delivery information: This is a stock item. Please allow 1-3 days for delivery.
Packaging: Frozen-Dry Ice