Coomassie brilliant blue G-250, Ultrapure

Synonyms: Xylene Brilliant Cyanine GC.I. 42655Hydrogen [4-[4-(p-ethoxyanilino)-4'-[ethyl(m-sulphonatobenzyl)amino]-2'-methylbenzhydrylene]-3-methylcyclohexa-2,5-dien-1-ylidene](ethyl)(m-sulphonatobenzyl)ammonium monosodium saltSodium 3-(((4-((4-((4-ethoxyphenyl)amino)phenyl)(4-(ethyl(3-sulphonatobenzyl)iminio)-2-methylcyclohexa-2,5-dien-1-ylidene)methyl)-3-methylphenyl)(ethyl)amino)methyl)benzenesulphonateServa Blue GHydrogen [4-[4-(p-ethoxyanilino)-4'-[ethyl(m-sulfonatobenzyl)amino]-2'-methylbenzhydrylene]-3-methylcyclohexa-2,5-dien-1-ylidene](ethyl)(m-sulfonatobenzyl)ammonium monosodium saltSodium 3-(((4-((4-((4-ethoxyphenyl)amino)phenyl)(4-(ethyl(3-sulfonatobenzyl)iminio)-2-methylcyclohexa-2,5-dien-1-ylidene)methyl)-3-methylphenyl)(ethyl)amino)methyl)benzenesulfonateAcid Blue 90Brilliant indocyanine GCoomassie Blue G 250Brilliant Blue GBrillant blue G

D202010
10497-494EA 109.98 USD
10497-494
Coomassie brilliant blue G-250, Ultrapure
Coomassie brilliant blue G-250
Formula: C₄₇H₄₈N₃NaO₇S₂
MW: 854.04 g/mol
MDL Number: MFCD00078482
CAS Number: 6104-58-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR