Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Rigol Digital Oscilloscope

Supplier: SAELIG COMPANY INC SE
DS4022 DS4054 DS4052 DS4014 DS4024 DS4034 DS4012
The dependable and versatile rigol digital oscilloscope features multiple channels and frequencies.

  • Bandwidths between 100 MHz and 500 MHz
  • Sample rates up to 4GSa/s
  • Up to four analog channels
  • Ultravision technology

The oscilloscope is ideal for a wide variety of markets including communications, defense, aerospace, industrial electronics, R and D, and educational fields.

The series features rigol's ultravision technology.

Delivery information: Resource CD and manual included.



 

ORDER

Expand All / Collapse All
Description Supplier No. VWR Catalog Number Unit Price Quantity
100 MHz, 2-Channel Digital Oscilloscope DS4012 470220-538 Each Retrieving
100 MHz, 4-Channel Digital Oscilloscope DS4014 470220-540 Each Retrieving
200 MHz, 2-Channel Digital Oscilloscope DS4022 470220-542 Each Retrieving
200 MHz, 4-Channel Digital Oscilloscope DS4024 470220-544 Each Retrieving
350 MHz, 4-Channel Digital Oscilloscope DS4034 470220-548 Each Retrieving
500 MHz, 2-Channel Digital Oscilloscope DS4052 470220-550 Each Retrieving
500 MHz, 4-Channel Digital Oscilloscope DS4054 470220-552 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Input Voltage 110V/220V

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty