NeuroPure™ E18 Rat Hypothalamus Astrocytes, Genlantis
Supplier: Genlantis
N800300
10497-554EA 859.8 USD
10497-554
NeuroPure™ E18 Rat Hypothalamus Astrocytes, Genlantis
Cells
NeuroPure Primary Rat Neurons are live neuronal tissues isolated from micro-surgically dissected regions of Sprague/Dawley rat.
  • Freshly isolated healthy neurons - not frozen
  • Ready to use - get your primary culture up and running within 1 hour
  • Pure neuronal cells - 99.9% glial cell free
  • Guaranteed quality and consistency
  • Available in seven popular cell types
These neurons are prepared fresh each week and shipped in a nutrient rich medium that keeps the cells alive for up to 6 days under refrigeration.