NeuroPure™ E18 Primary Mouse Midbrain Neuronal Cells, Genlantis
Supplier: Genlantis
N700201
10497-548EA 738.21 USD
10497-548
NeuroPure™ E18 Primary Mouse Midbrain Neuronal Cells, Genlantis
Cells
NeuroPure™ E18 primary mouse midbrain cells are live neurons provided as micro-surgically dissected regions of day 18 embryonic C57 mouse brain.
  • Mouse midbrain cells are prepared fresh each week
  • Shipped in a nutrient rich medium that keeps them alive for up to 14 days under refrigeration
  • Ideal for a wide variety of applications
Mouse midbrain cells are prepared fresh each week and shipped in a nutrient rich medium that keeps them alive for up to 14 days under refrigeration. Following simple dissociation steps, the NeuroPure cells can be quickly plated on almost any poly-lysine coated substrate using the provided NeuroPure plating medium. NeuroPure cells are ideal for a wide variety of applications including: transfection, pharmacology, electrophysiology, immunocytochemistry and neuronal development studies.
Delivery information: Includes day 18 Embryonic C57 Mouse Midbrain Cells 1 midbrain, NeuroPure™ Plating Medium 1 vial x 12 mL, NeuroPrep™ Medium 1 vial x 2.5 mL and NeuroPapain™ Enzyme 1 vial x 5 mg.