Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

NeuroPure™ E18 Primary Mouse Striatum Neuronal Cells, Genlantis

Supplier: Genlantis
N500201
NeuroPure™ E18 primary mouse striatum cells are live neurons provided as micro-surgically dissected regions of day 18 embryonic C57 mouse brain.

  • Mouse striatum cells are prepared fresh each week
  • Shipped in a nutrient rich medium that keeps them alive for up to 14 days under refrigeration
  • Can be quickly plated on almost any poly-lysine coated substrate

Mouse striatum cells are prepared fresh each week and shipped in a nutrient rich medium that keeps them alive for up to 14 days under refrigeration. Following simple dissociation steps, the NeuroPure cells can be quickly plated on almost any poly-lysine coated substrate using the provided NeuroPure plating medium. NeuroPure cells are ideal for a wide variety of applications including: transfection, pharmacology, electrophysiology, immunocytochemistry and neuronal development studies.

Delivery information: Includes day 18 Embryonic C57 Mouse Striatum Cells 1 striatum, NeuroPure™ Plating Medium 1 vial x 12 mL, NeuroPrep™ Medium 1 vial x 2.5 mL and NeuroPapain™ Enzyme 1 vial x 5 mg.



 

ORDER

Expand All / Collapse All
Description Size Cell Format Supplier No. VWR Catalog Number Unit Price Quantity
Day 18 Embryonic C57 Mouse Striatum Cells 1 vial x 2 mL Proliferating N500201 10497-540 Each Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty