Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Littmann® Cardiology IV™ Stethoscope, 3M™

Supplier: 3M Healthcare
MMM6158 MMM6157 MMM6156 MMM6155 MMM6154 MMM6153 MMM6152 MMM6151
The Cardiology IV™ Stethoscope is designed for clinicians who require outstanding acoustic performance combined with exceptional versatility

  • Outstanding acoustics with better audibility of high-frequency sounds
  • Hear high or low frequency sounds by slightly adjusting pressure on the chestpiece
  • Two sound paths in one tube eliminates the rubbing noise of traditional double tubes
  • Next-generation tubing lasts longer and resists stains

The innovative design provides a single-piece tunable diaphragm on each side of the chestpiece. Dual-lumen tubing combines two sounds paths in one tube, eliminating the rubbing noise that traditional double tubes create. Distinctive chalice-shaped chestpiece features tunable diaphragms on both sides to deliver outstanding acoustic performance, diagnostic versatility, and convenience for treating adult and pediatric patients.



 

ORDER

Expand All / Collapse All
Description Length Supplier No. VWR Catalog Number Unit Price Quantity
Cardiology IV™ Stethoscope, with Black Tubing 55.8 cm (22") MMM6151 75830-246 Each Retrieving Restricted
Cardiology IV™ Stethoscope, with Black Tubing 68.5 cm (27") MMM6152 75830-248 Each Retrieving Restricted
Cardiology IV™ Stethoscope, with Burgundy Tubing 68.5 cm (27") MMM6153 75830-322 Each Retrieving Restricted
Cardiology IV™ Stethoscope, with Caribbean Blue Tubing 68.5 cm (27") MMM6157 75830-250 Each Retrieving Restricted
Cardiology IV™ Stethoscope, with Hunter Green Tubing 68.5 cm (27") MMM6155 75830-252 Each Retrieving Restricted
Cardiology IV™ Stethoscope, with Navy Blue Tubing 68.5 cm (27") MMM6154 75830-316 Each Retrieving Restricted
Cardiology IV™ Stethoscope, with Plum Tubing 68.5 cm (27") MMM6156 75830-318 Each Retrieving Restricted
Cardiology IV™ Stethoscope, with Raspberry Tubing 68.5 cm (27") MMM6158 75830-320 Each Retrieving Restricted
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty