NeuroPure™ E18 Primary Rat Midbrain Neuronal Cells, Genlantis
Supplier: Genlantis
N700200
10497-546EA 859.8 USD
10497-546
NeuroPure™ E18 Primary Rat Midbrain Neuronal Cells, Genlantis
Cells
NeuroPure™ E18 primary rat midbrain cells are live neurons provided as micro-surgically dissected regions of day 18 embryonic sprague/dawley rat brain.
  • Freshly isolated healthy neurons
  • Ready to use with primary culture up and running within one hour
  • Pure neuronal cells - 99.9% glial cell free
Rat midbrain cells are prepared fresh each week and shipped in a nutrient rich medium that keeps them alive for up to 6 days under refrigeration. Following simple dissociation steps, the NeuroPure cells can be quickly plated on almost any poly-lysine coated substrate using the provided NeuroPure plating medium. NeuroPure cells are ideal for a wide variety of applications including: transfection, pharmacology, electrophysiology, immunocytochemistry and neuronal development studies.
Delivery information: Includes day 18 Embryonic Sprague/Dawley Rat Midbrain Cells 1 pair, NeuroPure™ Plating Medium 1 vial x 12 mL, NeuroPrep™ Medium 1 vial x 2.5 mL, NeuroPapain™ Enzyme 1 vial x 5 mg.