Ethidium bromide solution 0.636 mg/mL

Synonyms: 3,8-Diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide3,8-Diamino-5-ethyl-6-phenylphenanthridinium bromide

D100005
10497-492EA 125.84 USD
10497-492
Ethidium bromide solution 0.636 mg/mL
Ethidium bromide

Ethidium bromide dropper bottle contains high purity grade ethidium bromide at a concentration of 0.636 mg/mL in ultra pure water.


  • Dropper bottles are shatter-resistant and non-breakable
  • Offers convenience and safety
  • Ideal for staining DNA in agarose electrophoresis gels
  • Sufficient for about 125 x 50 mL gels


The Ethidium Bromide Dropper Bottle contains high purity grade ethidium bromide at a concentration of 0.636 mg/ml in ultra pure water. Provided in a 5 mL non-breakable dropper bottle, this product is ideal for staining DNA in agarose electrophoresis gels. The dropper format allows researchers to minimize exposure to ethidium bromide and offers a convenient, consistent method for all nucleic acid staining applications. The
Ethidium Bromide Dropper Bottle is sufficient for about 125 x 50 mL gels.

Formula: C₂₁H₂₀BrN₃
MW: 394.31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011724
CAS Number: 1239-45-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR