ATP Test Kits, Modern Water Inc
Supplier: Modern Water
QGA-25C QGA-100C QG21W-50C QGOM-25C DSA-25C QGOM-100C
75812-686BX 1936.65 USD
75812-686 75812-684 75812-688 75812-690 75812-694 75812-692
ATP Test Kits, Modern Water Inc
Analyzers Elemental Analysers
ATP Test Kits provide fast, complete and accurate insight into microbial activity.
  • Fast, complete and accurate insight into microbial activity
  • Highly sensitive
  • Measure any filterable water sample
ATP Test Kits provide fast, complete and accurate insight into microbial activity
Organic Modified test kit is specifically designed to measure total microorganisms in organic fluids. Wastewater tests used for biological wastewater treatment.
Deposit & Surface analysis used for slimes, biofilms, coupons and biological filter media. Luminometer is required.
Provides highly-sensitive and interference free measurement of total living microorganisms in nearly any filterable water sample.