Would you like to log in again?
X-Gal Staining™ Assay Kit, Genlantis
Supplier: Genlantis
A10300K
The X-Gal staining assay kit is designed for sensitive and low-background in situ detection of cells transfected with vectors carrying the lacZ gene.
  • Sensitive detection of β-galactosidase activity in situ
  • Results in less than 24 hours post-transfection
  • Easy visualization of positive cells
  • Direct assay of transfections in cell culture dishes or in tissues in vivo
This simple and rapid assay is suitable for expression studies in cultured cells and tissues transfected in vivo. The β-galactosidase enzyme catalyzes the hydrolysis of X-gal (5-bromo-3-indoyl-β-D-galactopyranoside), a β-galactoside. Following fixation and incubation with the X-gal substrate, cells transfected with a β-galactosidase-expressing plasmid will appear blue. These blue cells can be easily visualized by microscopy.
Delivery information: Kit includes Fixing buffer 1 vial x 125 mL, 10X PBS 1 vial x 75 mL, Staining Buffer 1 vial x 125 mL and 25X X-Gal Stock 4 vials x 1 mL.
ORDER
Expand All / Collapse All
Description Size Supplier No. VWR Catalog Number Unit Price Quantity
X-Gal Staining Assay Kit (50 Assays in 60 mm dishes) Kit A10300K 10497-480 Each Retrieving
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Q and A
Product Questions & Answers
Sort by: Most Helpful | Newest | Oldest
Items Per Page: 5  10  20 
2  3  4  Next
Can't find an answer to your question?  Ask us a question
Ask us a question:
Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question
Q&A Guidelines & Tips
If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.
When writing your question or answer, please follow these guidelines:
  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English
All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s
We reserve the right not to post your question if it contains any of the following
  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty