Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

DeltaTox II Analyzer with Case, Modern Water Inc

Supplier: MODERN WATER INC.
AZF848523 AZF686011 AZF686005 AZF686004
DeltaTox® II is a simple, rapid, portable water quality test system, designed for acute toxicity screening and total microbial contamination using ATP.

  • Simple, rapid, portable
  • Bioluminescence technology
  • Total microbial contamination using ATP

DeltaTox® II is a simple, rapid, portable water quality test system, designed for acute toxicity screening and total microbial contamination using ATP. DeltaTox® II uses bioluminescence technology to screen for contamination in instances of drinking water emergencies and chemical spills into water systems.

Certifications: CE compliant.

Ordering information: Also Required 50 Vials of SOLO Reagent, 1 L of Diluent, 50 mL Osmotic Adjusting Solution, 10-100 uL Adjustable Volume Pipet, 100-100 uL Adjustable Volume Pipet, 10-100 uL Tips (96), 100-1000 uL Tips (100), Disposable Glass Cuvettes (400), Cuvette Racks (5).



 

ORDER

Expand All / Collapse All
Description Size Supplier No. VWR Catalog Number Unit Price Quantity
DeltaTox II Analyzer with Case AZF848523 75812-681 Each Retrieving
Microtox Diluent 1 L AZF686011 75812-670 Each Retrieving
Disposable Glass Cuvettes Case of 1000 AZF686004 75812-678 Case of 1 Retrieving
Disposable Glass Cuvettes Case of 400 AZF686005 75812-676 Case of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

ACCESSORIES

Expand All / Collapse All

Accessories
Description Size Details Supplier No. VWR Catalog Number Unit Price Quantity
Microtox SOLO Reagent 50 vials Storage Temperature (–25° C) AZF686300 75812-682 Box of 1 Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Microtox Osmotic Adjusting Solution 50 mL AZF686019 75812-672 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty