Anti-LILRB3 Mouse Monoclonal Antibody (Alexa Fluor 488) [clone: 222821]

FAB1806G-025
103251-828EA 153.85 USD
103251-828
Anti-LILRB3 Mouse Monoclonal Antibody (Alexa Fluor 488) [clone: 222821]
Type: Primary
Antigen: LILRB3
Clonality: Monoclonal
Clone: 222821
Conjugation: Alexa Fluor 488
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR