Anti-IL17RA Mouse Monoclonal Antibody (Phycoerythrin) [clone: 133617]

Supplier: Novus Biologicals
FAB177P-025 FAB177P-100
103251-798EA 533.63 USD
103251-798 103251-796
Anti-IL17RA Mouse Monoclonal Antibody (Phycoerythrin) [clone: 133617]
Antibodies
Type: Primary
Antigen: IL17RA
Clonality: Monoclonal
Clone: 133617
Conjugation: Phycoerythrin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR